Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iminodiacetonitrile, 95%

Catalog No. AAB236600E
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 628-87-5 | C4H5N3 | 95.11 g/mol

Iminodiacetonitrile is used as a chemical intermediate. It is also used in the preparation of iminodiacetic acid. Further, it is used to study the interference of nitriles in cyanide determination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iminodiacetonitrile is used as a chemical intermediate. It is also used in the preparation of iminodiacetic acid. Further, it is used to study the interference of nitriles in cyanide determination.

Solubility
Soluble in water, acetone and hot methanol.

Notes
Incompatible with strong acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-87-5
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.11
MDL Number MFCD00001887
InChI Key BSRDNMMLQYNQQD-UHFFFAOYSA-N
Synonym iminodiacetonitrile, 2,2'-azanediyldiacetonitrile, acetonitrile, 2,2'-iminobis, 1,1'-imidodiacetonitrile, acetonitrile, iminodi, 2,2'-iminodiacetonitrile, 2,2'-iminobisacetonitrile, bis cyanomethyl amine, di cianometil ammina, 2-cyanomethyl amino acetonitrile
PubChem CID 69413
ChEBI CHEBI:71189
IUPAC Name 2-(cyanomethylamino)acetonitrile
SMILES N#CCNCC#N

Specifications

Melting Point 75°C to 79°C
Quantity 2500 g
Beilstein 773803
Solubility Information Soluble in water,acetone and hot methanol.
Formula Weight 95.11
Percent Purity 95%
Chemical Name or Material Iminodiacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 211-058-3
RTECSNumber AM0650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.