Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Imipramine hydrochloride
GREENER_CHOICESDP

Catalog No. 784150 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
78-415-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 78-415-0 Supplier Tocris Bioscience™ Supplier No. 7841/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent inhibitor of 5-HT and noradrenalin transporters; promotes immunostimulatory tumor microenvironment

Imipramine hydrochloride is a potent tricyclic antidepressant; inhibits 5-HT and noradrenalin transporters (Ki values = 1.3-20 nM and 20-142 nM respectively). Increases autophagy, both alone and in combination with VEGF inhibitors or Temozolomide (Cat. No. 2706), to reduce the progression of glioma and slow the proliferation of other tumors. Reprograms tumor-associated macrophages from immunosuppressive to immunostimulatory via histamine receptor inhibition.

Specifications

Chemical Name or Material 5-(3-Dimethylaminopropyl)-10,11-dihydro-5H-dibenz[b,f]azepine hydrochloride
CAS 113-52-0
Quantity 50 mg
Target Autophagy-related Compounds
Molecular Formula C19H24N2.HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.