Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imipramine Hydrochloride, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18603433
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle

IM120, 113-52-0, C19H24N2·HCl

Spectrum™ Chemical Imipramine Hydrochloride, USP is used as an antidepressant and for the treatment of nighttime bed-wetting (enuresis). All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 113-52-0
Molecular Formula C19H25ClN2
Molecular Weight (g/mol) 316.87
InChI Key XZZXIYZZBJDEEP-UHFFFAOYSA-N
IUPAC Name hydrogen (3-{2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-yl}propyl)dimethylamine chloride
SMILES [H+].[Cl-].CN(C)CCCN1C2=CC=CC=C2CCC2=CC=CC=C12

Specifications

CAS Max % 100%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 98 to 102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.