Learn More
Cytiva Immobiline™ DryPlate for IEF
Catalog No. 45001618 Shop All Cytiva Products
Change view
Click to view available options
Medium range; pH 4-7
Catalog No.
45-001-618 Medium range; pH 4-7
1 options
Catalog No. 45-001-618 Supplier Cytiva Supplier No. 80112828
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Obtain reproducible IEF separations with dry, precast gels containing immobilized pH gradients
Immobiline DryPlate precast gels contain preformed, immobilized, non-drifting pH gradients. The gels are rehydrated to a thickness of 0.5mm in the reswelling cassette accessory before use.
  • Medium range covering 3 pH units
  • Handle mechanically stable, plastic-backed gels with confidence
  • Immobiline technology produces an absolutely stable pH gradient, allowing extremely high voltages and high resolution over narrow pH ranges, even for low mobility proteins that require long focusing times
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) IEF
Type Precast Gel