Learn More
Description
Choose from a comprehensive range of overlapping Immobiline DryStrip gels that cover narrow (1 pH unit), medium (3 to 5 pH units), and broad (8 pH units) pH ranges. The pH gradients at the extreme ends of the pH scale are nonlinear to distribute the proteins evenly over the gel length to obtain maximum resolution.
Five narrow pH range (1 pH unit), 24-cm IPG strips deliver high resolution and protein loading capacity, which results in well separated spots with high protein content after the second-dimension separation
- Select the broad pH 3 to 11 Non-Linear (NL) gradient Immobiline DryStrip gel for fast and efficient screening to gain a broad overview of total protein disribution
- For more detailed studies or when using greater protein loads, choose from four new overlapping strips covering the pH 3 to 11 range
- Higher resolution can be obtained using narrow-range (1 pH unit) Immobiline DryStrip gels
- Use matching IPG Buffers for optimized running conditions
- Use IPG strips directly after reswelling without further manipulation
- Bar coded strips for convenient sample and batch tracking
- Immobiline DryStrip gels are rehydrated with a matching IPG Buffer prior to use
- Samples can be included directly in the rehydration solution for efficient loading of dilute samples or for loading larger amounts of protein
- Samples can also be loaded after rehydration
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Immobiline DryStrip gels (IPG strips) are used for isoelectric focusing (IEF), run as the first dimension of 2-D electrophoresis or as a separate application. |
| Color | Colorless |
| Format | Precast Gel |
| pH Range | 3 to 10 |
| Separation Type | Isoelectric Focusing |
| Form | Dried |
| For Use With (Application) | Industrial applications: analytical chemistry, research |
| Gradient | Non-linear |
| Storage Requirements | −20°C |
| Thickness (English) | 0.19 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
