Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Immobiline™ DryStrip Gels

Catalog No. 45000406 Shop All Cytiva Products
Change view
Click to view available options
pH Range:
3 to 10
3 to 11
4 to 7
6 to 11
6 to 9
Gradient:
Linear
Non-linear
Length (Metric):
7 cm
11 cm
13 cm
18 cm
24 cm
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range Gradient Length (Metric)
45-000-406 3 to 11 Non-linear 24 cm
45-000-213 3 to 10 Non-linear 18 cm
45-000-362 6 to 11 Linear 18 cm
45-000-361 6 to 11 Linear 13 cm
45-000-359 6 to 11 Linear 7 cm
45-000-379 6 to 9 Linear 24 cm
45-000-377 3 to 10 Non-linear 24 cm
45-000-402 3 to 11 Non-linear 7 cm
45-000-404 3 to 11 Non-linear 13 cm
45-000-211 4 to 7 Linear 18 cm
45-000-212 3 to 10 Linear 18 cm
45-000-347 3 to 10 Linear 7 cm
45-000-348 3 to 10 Non-linear 7 cm
45-000-346 4 to 7 Linear 7 cm
45-000-349 4 to 7 Linear 13 cm
45-000-350 3 to 10 Linear 13 cm
45-000-351 3 to 10 Non-linear 13 cm
45-000-360 6 to 11 Linear 11 cm
45-000-378 4 to 7 Linear 24 cm
45-000-376 3 to 10 Linear 24 cm
45-000-405 3 to 11 Non-linear 18 cm
45-000-403 3 to 11 Non-linear 11 cm
45-000-428 4 to 7 Linear 11 cm
45-000-429 3 to 10 Linear 11 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
Catalog No. 45-000-406 Supplier Cytiva Supplier No. 17600377
Add to Cart
Add to Cart

Represent a comprehensive selection of narrow-, medium- and wide-range IPG strips. Cytiva Life Sciences™ Immobiline™ DryStrip gels (IPG strips) are used for isoelectric focusing (IEF), run as the first dimension of 2-D electrophoresis or as a separate application.

Choose from a comprehensive range of overlapping Immobiline DryStrip gels that cover narrow (1 pH unit), medium (3 to 5 pH units), and broad (8 pH units) pH ranges. The pH gradients at the extreme ends of the pH scale are nonlinear to distribute the proteins evenly over the gel length to obtain maximum resolution.

Five narrow pH range (1 pH unit), 24-cm IPG strips deliver high resolution and protein loading capacity, which results in well separated spots with high protein content after the second-dimension separation

  • Select the broad pH 3 to 11 Non-Linear (NL) gradient Immobiline DryStrip gel for fast and efficient screening to gain a broad overview of total protein disribution
  • For more detailed studies or when using greater protein loads, choose from four new overlapping strips covering the pH 3 to 11 range
  • Higher resolution can be obtained using narrow-range (1 pH unit) Immobiline DryStrip gels
  • Use matching IPG Buffers for optimized running conditions
  • Use IPG strips directly after reswelling without further manipulation
  • Bar coded strips for convenient sample and batch tracking

  • Immobiline DryStrip gels are rehydrated with a matching IPG Buffer prior to use
  • Samples can be included directly in the rehydration solution for efficient loading of dilute samples or for loading larger amounts of protein
  • Samples can also be loaded after rehydration

TRUSTED_SUSTAINABILITY

Specifications

Description Immobiline DryStrip gels (IPG strips) are used for isoelectric focusing (IEF), run as the first dimension of 2-D electrophoresis or as a separate application.
Color Colorless
Format Precast Gel
pH Range 3 to 11
Separation Type Isoelectric Focusing
Form Dried
For Use With (Application) Industrial applications: analytical chemistry, research
Gradient Non-linear
Storage Requirements −20°C
Thickness (English) 0.19 in.
Thickness (Metric) 0.05 cm
Length (Metric) 24 cm
Width (English) 0.118 in.
Width (Metric) 0.3 cm
Product Type Precast Gel
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.