Description
Run up to 12 Immobiline DryStrip strips simultaneously with up to 100μL on each Immobiline DryStrip.
- The instruction manual presents all necessary information for the entire method for horizontal 2-D electrophoresis. By appropriate adaptations, the SDS electrophoresis can also be performed using vertical electrophoresis equipment
- Immobiline DryStrip kit facilitates sample application and running and equilibration of Immobiline DryStrip for the first dimension in 2-D (two-dimensional) electrophoresis
- Isoelectric focusing with an immobilized pH gradient (IPG), makes true isoelectric focusing possible and significantly improves the reproducibility of the spot distribution along the pH gradient axis of 2-D maps
- Individual components are also available separately
Specifications
Specifications
| For Use With (Application) | First-dimension IEF |
| For Use With (Equipment) | Multiphor II Electrophoresis System |
| Type | Immobiline DryStrip Kit |
| Includes | Anode, cathode, sample cup bar, tray and electrode holder, 100μL sample cups, DryStrip aligners, IEF electrode strips and sample application pieces |