Learn More
Cytiva Immobiline™ Sample Application Strips
Catalog No. 45001796 Shop All Cytiva Products
Change view
Click to view available options
For 26 SDS samples; 40μL
Catalog No.
45-001-796 For 26 SDS samples; 40μL
1 options
Catalog No. 45-001-796 Supplier Cytiva Supplier No. 18100274
Offer improved gel performance with faster and more stable gradients, fewer artifacts, and easier zymogram assays
  • For use with SDS and immobiline samples
  • Silicone applicator strip with sample holes to load samples onto Isoelectric focussing (IEF) Gels
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) 26 SDS sample
Type SDS Sample Application Strip
Quantity 5