Learn More
Thermo Scientific™ Immobilized Anti-TMT™ Antibody Resin, 6 mL PROMO
Description
Tandem mass tag (TMT) reagents are used to chemically label proteins in different samples for multiplexed quantitation using tandem mass spectrometry. Immobilized Anti-TMT Resin contains an agarose-immobilized anti-TMT monoclonal antibody and is effective for capture and enrichment of peptides or proteins labeled using any TMT reagent. This helps simplify otherwise complex samples before LC-MS/MS analysis, especially when the targets for labeling are rare (e.g., cysteine sulfhydryls). Enrichment of labeled peptides by this method results in fewer peptides identified per protein, but can significantly increase the number of unique proteins identified, making it easier to detect and measure low-abundance proteins.
Immobilized Anti-TMT Resin uses our highly specific Anti-TMT Antibody (Cat. No. 90075) to bind peptides labeled with TMT reagents. The antibody is specific for the mass reporter region of the TMT reagents, which allows for enrichment of TMT-labeled peptides regardless of peptide labeling chemistry (e.g., amine-reactive, sulfhydryl-reactive). TMT Elution Buffer is a volatile neutral buffer that competitively elutes captured TMT-labeled peptides. When used in combination with Immobilized Anti-TMT Resin, TMT Elution Buffer provides higher specific enrichment of TMT-labeled peptides compared to acidic or basic elution buffers.
Enrichment features include:
- Specific—uses specific, high-affinity antibody for the TMT structure (Kd >100 pM)
- Compatible—effective for all varieties of TMT reagents (amine-reactive or cysteine-reactive)
- Versatile—use resin with TMT Elution Buffer for optimal sample enrichment
Specifications
Specifications
Product Type | Immobilized Anti-TMT Antibody Resin |
Product Line | TMT™ |
Content And Storage | Upon receipt store all products at 4°C. Product shipped with an ice pack. |
Label or Dye | Tandem Mass Tag™ (TMT™) Labels |
Starting Material | Cell Lysate |
Detection Method | Mass Spectrometry |
Workflow Step | Protein Labeling |
Quantity | 6 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.