Learn More
Thermo Scientific™ Immobilized Ficin (Agarose Resin) PROMO
Description
Ficin, which is similar to papain, is a nonspecific, sulfhydryl protease isolated from fig latex. The molecular weight of Ficin is approximately 25,000 and it is effective at pH 4-9.5 with an optimum pH of 6.5. When IgG molecules are incubated with ficin in the presence of cysteine, one or more peptide bonds in the hinge region are split, producing either Fab fragments or F(ab')2 fragments. Mouse IgG1 is notably difficult to digest into functional Fab or F(ab')2 fragments using papain or pepsin, however, yields and immunoreactivity are better when digesting mouse IgG1 with ficin. Immobilized Ficin was specifically designed for cleavage of mouse IgG1 into or Fab or F(ab')2 fragments, and is advantageous because it virtually eliminates autolysis, eliminates contamination of a sample with the protease and allows control of the digestion by removing the ficin or controlling the flow of sample over an immobilized ficin column.
Highlights:
- Can generate both Fab and F(ab')2 fragments from mouse IgG1
- Reaction can be easily controlled
- Antibody fragments are free of autodigestion products
- Ficin contamination into antibody fragments is eliminated
Specifications
Specifications
Content And Storage | Content: 5 mL of settled resin. Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2°C to 8°C. |
Form | Liquid Suspension |
Quantity | 5 mL |
Product Type | Ficin |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.