Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Immobilized Trypsin, TPCK Treated (Agarose Resin) PROMO
Click to view available options
:
Thermo Scientific™ Immobilized Trypsin, TPCK Treated (Agarose Resin)
Description
Highlights:
- Immobilized TPCK Trypsin eliminates trypsin contamination in digests
- TPCK inhibits chymotrypsin activity
Specifications
Specifications
Content And Storage | Contents: Immobilized TPCK Trypsin, 2 mL of settled gel supplied as a 50% slurry containing glycerol and 0.05% sodium azide. Storage: Upon receipt Store at 4°C. Product is shipped at ambient temperature. |
Form | Liquid Suspension |
Quantity | 2 mL |
Product Type | Trypsin |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction