Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Immobilon-E NOW Bundle

Catalog No. NWKITIE Shop All MilliporeSigma Products
Change view
Click to view available options
Code:
IEVH
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Code
NWKITIE IEVH
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NWKITIE Supplier MilliporeSigma™ Supplier No. NWKITIE
Add to Cart
Add to Cart

Immobilon-E is the first PVDF membrane for Western blotting that can be wetted with with aqueous transfer buffer, eliminating the alcohol pre-wet step required with other PVDF membranes. For chemiluminescence or chromogenic detection.

  • Includes the Immobilon NOW Dispenser and two 8.5cm x 10m rolls of Immobilon-E transfer membrane
  • System offers 1-cut convenience for standard mini-blots (7 x 8.5cm) and midi-blots (8.5 x 13.5cm)
  • 45 μm pore size recommended for most Western blotting applications, especially proteins greater than 20 kDa
  • Detection method: Chemiluminescence Chromogenic Radioactive
  • Compatible stains:
    • Coomassie brilliant blue
    • Amido black
    • India ink
    • Ponceau-S red
    • Colloidal gold
    • Transillumination
    • Sypro ruby
  • Primary Binding Mechanisms: Electrostatic, Hydrophobic
  • Easily wetted with water
  • Filter dimensions; 8.5 cm x 10 m
  • Chemistry: Polyvinylidene Fluoride (PVDF)
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blotting; Immunoblotting
Color White
Code IEVH
Surface Plain
Material PVDF
Pore Size 0.45 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.