Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IMMP1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575712
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575712 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575712 Supplier Invitrogen™ Supplier No. PA575712
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

The mitochondrial inner membrane peptidase (IMP) complex generates mature, active proteins in the mitochondrial intermembrane space by proteolytically removing the mitochondrial targeting presequence of nuclear-encoded proteins. IMP1 and IMP2 (IMMP2L; MIM 605977) are the catalytic subunits of the IMP complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IMMP1L
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.2
Gene IMMP1L
Gene Accession No. Q96LU5, Q9CQU8
Gene Alias 1500034J20Rik; 2610528O17Rik; AU042386; IMMP1; Immp1l; IMP1; IMP1 inner mitochondrial membrane peptidase-like; IMP1 inner mitochondrial membrane peptidase-like (S. cerevisiae); IMP1-LIKE; IMP1-like protein; inner mitochondrial membrane peptidase subunit 1; mitochondrial inner membrane protease subunit 1
Gene Symbols IMMP1L
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human IMMP1L.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196294, 66541, 691145
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.