Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IMMP1L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106945
Description
Antibody detects endogenous levels of total IMMP1L.
The mitochondrial inner membrane peptidase (IMP) complex generates mature, active proteins in the mitochondrial intermembrane space by proteolytically removing the mitochondrial targeting presequence of nuclear-encoded proteins. IMP1 and IMP2 (IMMP2L; MIM 605977) are the catalytic subunits of the IMP complex.
Specifications
IMMP1L | |
Polyclonal | |
Unconjugated | |
IMMP1L | |
1500034J20Rik; 2610528O17Rik; AU042386; IMMP1; Immp1l; IMP1; IMP1 inner mitochondrial membrane peptidase-like; IMP1 inner mitochondrial membrane peptidase-like (S. cerevisiae); IMP1-LIKE; IMP1-like protein; inner mitochondrial membrane peptidase subunit 1; mitochondrial inner membrane protease subunit 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
196294, 66541 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96LU5, Q9CQU8 | |
IMMP1L | |
A synthesized peptide derived from human IMMP1L(Accession Q96LU5), corresponding to amino acid residues F102-G152. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction