Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IMMP2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555387
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555387 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555387 Supplier Invitrogen™ Supplier No. PA555387
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NRYVKVPRGH IWVEGDHHGH SFDSNSFGPV SLGLLHAHAT HILWPPERWQ KLESVLPPER LPVQREEE Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 26%.

IMP2L complex generates mature, active proteins in the mitochondrial intermembrane space by proteolytically removing the mitochondrial targeting presequence of nuclear-encoded proteins. IMP1 and IMP2 are the catalytic subunits of the IMP complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IMMP2L
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IMMP2L
Gene Accession No. Q96T52
Gene Alias AI853880; IMMP2L; IMMP2L-IT1; IMP2; IMP2 inner mitochondrial membrane peptidase-like; IMP2 inner mitochondrial membrane peptidase-like (S. cerevisiae); IMP2-LIKE; IMP2-like protein; inner mitochondrial membrane peptidase 2 like; inner mitochondrial membrane peptidase subunit 2; mitochondrial inner membrane protease subunit 2
Gene Symbols IMMP2L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IMMP2L. Recombinant protein control fragment (Product #RP-92828).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83943
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.