Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Immuno Breakable Modules Clear
GSA_VA

Catalog No. p-4525323
Click to view available options
Surface Treatment:
MaxiSorp
PolySorp
Universal Binding
Well Shape:
Round, C Bottom
Round, Flat Bottom (F-well)
No. per Pack:
5
10
50
Product Type:
Microplate Strip
Strip Plate
This item is not returnable. View return policy
This item is not returnable. View return policy

Choose from four surfaces and a variety of wells and configurations with these immuno breakable modules, which offer an optimal choice of products for the specific assay.

Choose from four surfaces and a variety of wells and configurations with Thermo Scientific™ Immuno Breakable Modules, which offer reliability, well-to-well reproducibility and an optimal choice of products for the specific assay.

Choose between LockWell, BreakApart and Breakable 1x8 modules in standard 96-well frames.

Features:

  • For solid-phase immunoassays, e.g. ELISA
  • Choice of four surfaces for optimal binding
  • Free-standing modules are easy to handle
  • Alphanumeric system for easy orientation
  • Fits standard equipment
  • High optical quality for reliable results
  • Polystyrene

Choice of Surface:

  • PolySorp™, Immulon™ 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
  • Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
  • MaxiSorp™, Immulon 4 HBX, and Enhanced Binding (EB) are hydrophilic and ideal for antibody sandwich assays
  • MultiSorp™ are very hydrophilic and work well for glycans and water-soluble proteins

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Breakapart Breakable Strips (LockWell)
Binding Property Hydrophobic
Certified No
Format Strip Plate
Lid Without Lid
Description High optical quality for reliable results
Detection Method Colorimetric
Array 8 x 12
Packaging Inner packs of 5
Color Clear
Product Type Strip Plate
Material Polystyrene
Volume (Metric) Well 350 μL
No. of Wells 96
Volume (Metric) Working 250 μL
Sterility Non-sterile
Surface Treatment PolySorp
Well Shape Round, C Bottom
For Use With (Application) For adsorption of hydrophobic molecules
No. per Case 60
No. per Pack 5
Plate Blocking None
Quantity 60/Cs.
Target Molecule Lipids
Well Design C8
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.