Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Immuno Standard Modules White and Black

Catalog No. 12565275
Click to view available options
Color:
Black
White
Volume (Metric) Well:
350 μL
380 μL
400 μL
Surface Treatment:
Hydrophobic
MaxiSorp
PolySorp
This item is not returnable. View return policy
This item is not returnable. View return policy

Conduct fluorescence- and luminescence-based immunoassays and binding assays using these immuno standard modules white and black.

Conduct fluorescence- and luminescence-based immunoassays and binding assays using Thermo Scientific™ Immuno Standard Modules White and Black.

Features:

  • Available in 8-, 12- or 16-well strips
  • Framed or unframed for choice of optimal assay
  • Flat-bottom or C-shaped wells
  • Choice of three surface types for optimal binding
  • White format for highest possible signal reflection with low background fluorescence; recommended for epifluorescence
  • Black format to reduce backscatter light, which may be encountered in epifluorescence
  • Transparent or clear format for use in time-resolved fluorescence applications only

Choice of Surface:

  • Thermo Scientific™ PolySorp™ and Microlite™ 1+ are for adsorption of hydrophobic molecules.
  • Microlite 2+ is slightly hydrophilic and binds a diverse range of biomolecules.
  • Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assay.

TRUSTED_SUSTAINABILITY

Specifications

Breakapart No
Binding Property Hydrophobic
Certified No
Format Standard
Detection Method Fluorescence
Array 8 x 12
Packaging 60 Cases
Binding Type Adsorption
Color Black
Product Type Microplate Strip
Material Polystyrene
Volume (Metric) Well 400 μL
No. of Wells 96
Volume (Metric) Working 350 μL
Sterility Non-sterile
Surface Treatment PolySorp
Well Shape Round, Flat Bottom (F-well)
For Use With (Application) Fluorescence
No. per Case 60
No. per Pack 5
Plate Blocking None
Quantity 60/Cs.
Target Molecule Lipids
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.