Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Immuno Standard Modules White and Black
GSA_VA

Catalog No. p-4525215
Click to view available options
Color:
Black
White
Volume (Metric) Well:
350 μL
380 μL
400 μL
Surface Treatment:
Hydrophobic
MaxiSorp
PolySorp
This item is not returnable. View return policy

Catalog No. 12-565-289A

Supplier: Thermo Scientific™ 437591

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Conduct fluorescence- and luminescence-based immunoassays and binding assays using these immuno standard modules white and black.

Conduct fluorescence- and luminescence-based immunoassays and binding assays using Thermo Scientific™ Immuno Standard Modules White and Black.

Features:

  • Available in 8-, 12- or 16-well strips
  • Framed or unframed for choice of optimal assay
  • Flat-bottom or C-shaped wells
  • Choice of three surface types for optimal binding
  • White format for highest possible signal reflection with low background fluorescence; recommended for epifluorescence
  • Black format to reduce backscatter light, which may be encountered in epifluorescence
  • Transparent or clear format for use in time-resolved fluorescence applications only

Choice of Surface:

  • Thermo Scientific™ PolySorp™ and Microlite™ 1+ are for adsorption of hydrophobic molecules.
  • Microlite 2+ is slightly hydrophilic and binds a diverse range of biomolecules.
  • Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assay.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Breakapart No
Binding Property Hydrophilic
Certified Yes
Format Standard
Lid Without Lid
Detection Method Chemiluminescence
Array 8 x 12
Packaging 60 Cases
Binding Type Adsorption
Color White
Product Type Microplate Strip
Material Polystyrene
Volume (Metric) Well 350 μL
No. of Wells 96
Volume (Metric) Working 250 μL
Surface Treatment MaxiSorp
Well Shape Round, C Bottom
For Use With (Application) Fluorescence and Luminescence
No. per Case 60
No. per Pack 5
Plate Blocking None
Target Molecule Antibodies (Glycoproteins)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.