Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ImmunoCAP™ Epidermal and Animal Protein Allergens

Catalog No. 14411001P
Change view
Click to view available options
Quantity:
10 Tests
16 Tests
Code:
Re206
Re209
Re211
Re214
Re215
Re217
Re218
Re219
e1
e201
e3
e4
e5
e6
e7
e70
e71
e72
e73
e74
e75
e76
e77
e78
e80
e81
e82
e83
e84
e85
e86
e87
e88
e89
Common Name:
Budgerigar droppings
Budgerigar feathers
Canary bird feathers
Cat dander
Chicken droppings
Chicken feathers
Chicken, serum proteins
Cow dander
Dog dander
Duck feathers
Ferret epithelium
Finch feathers
Gerbil epithelium
Goat epithelium
Goose feathers
Guinea pig epithelium
Hamster epithelium
Horse dander
Mouse epithelium
Mouse epithelium, serum proteins and urine proteins
Mouse serum proteins
Mouse urine proteins
Pigeon droppings
Pigeon feathers
Rabbit epithelium
Rabbit, serum proteins
Rabbit, urine proteins
Rat epithelium
Rat epithelium, serum proteins and urine proteins
Rat serum proteins
Rat urine proteins
Sheep epithelium
Swine epithelium
Turkey feathers
Regulatory Status:
ASR
FDA Cleared
34 product options available for selection
Product selection table with 34 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Code Common Name Regulatory Status
14-411-001P 16 Tests e5 Dog dander FDA Cleared
14-410-901P 16 Tests e1 Cat dander FDA Cleared
14-412-201P 16 Tests e3 Horse dander FDA Cleared
14-416-601P 16 Tests e4 Cow dander FDA Cleared
14-416-701P 16 Tests e6 Guinea pig epithelium FDA Cleared
14-416-901P 16 Tests e70 Goose feathers FDA Cleared
14-417-001P 10 Tests e85 Chicken feathers FDA Cleared
14-417-101P 10 Tests e86 Duck feathers FDA Cleared
14-430-401P 10 Tests e78 Budgerigar feathers FDA Cleared
14-430-801P 10 Tests e82 Rabbit epithelium FDA Cleared
14-430-301P 10 Tests e77 Budgerigar droppings FDA Cleared
14-430-701P 10 Tests e81 Sheep epithelium FDA Cleared
14-430-901P 10 Tests e83 Swine epithelium FDA Cleared
14-430-601P 10 Tests e80 Goat epithelium FDA Cleared
14-431-001P 10 Tests e84 Hamster epithelium FDA Cleared
14-432-601P 10 Tests e87 Rat epithelium, serum proteins and urine proteins FDA Cleared
14-432-701P 10 Tests e88 Mouse epithelium, serum proteins and urine proteins FDA Cleared
14-435-601P 10 Tests e89 Turkey feathers FDA Cleared
14-439-401P 10 Tests e71 Mouse epithelium FDA Cleared
14-439-501P 10 Tests e72 Mouse urine proteins FDA Cleared
14-439-701P 10 Tests e74 Rat urine proteins FDA Cleared
14-439-601P 10 Tests e73 Rat epithelium FDA Cleared
14-439-901P 10 Tests e76 Mouse serum proteins FDA Cleared
14-439-801P 10 Tests e75 Rat serum proteins FDA Cleared
14-482-401P 10 Tests e201 Canary bird feathers FDA Cleared
14-501-310P 10 Tests Re214 Finch feathers ASR
14-501-510P 10 Tests Re209 Gerbil epithelium ASR
14-503-410P 10 Tests Re211 Rabbit, urine proteins ASR
14-503-210P 10 Tests Re215 Pigeon feathers ASR
14-503-310P 10 Tests Re206 Rabbit, serum proteins ASR
14-515-710P 10 Tests Re217 Ferret epithelium ASR
14-523-010P 10 Tests Re219 Chicken, serum proteins ASR
14-522-910P 10 Tests Re218 Chicken droppings ASR
14-416-801P 16 Tests e7 Pigeon droppings FDA Cleared
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 34 options available.
34 missing translation for 'options'
This item is not returnable. View return policy
Catalog No. 14-411-001P missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' 14411001
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measure specific IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.

Quantify specific IgE antibodies with ImmunoCAP™ Epidermal and Animal Protein Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.

ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.
TRUSTED_SUSTAINABILITY

Specifications

Regulatory Status FDA Cleared
Code Type Animal Protein
Common Name Dog dander
Quantity 16 Tests
Code e5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.