Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ImmunoCAP™ Food Allergen Mixes

Catalog No. 14485001P
Click to view available options
Quantity:
10 Tests
16 Tests
Code:
Rfx17
Rfx21
Rfx22
Rfx70
Rfx71
Rfx72
Rfx74
fx1
fx13
fx14
fx15
fx16
fx18
fx2
fx20
fx24
fx25
fx3
fx5
fx7
fx73
Common Name:
f1, f2, f3, f4, f13, f14, Egg white, Milk, Fish, Wheat, Peanut, Soybean
f10, f45, f47, f85, Sesame seed, Yeast, Garlic, Celery
f12, f13, f14, Pea, Peanut, Soybean
f12, f15, f31, f35, Pea, White bean, Carrot, Potato
f13, f17, f18, f20, f36, Peanut, Hazel nut, Brazil nut, Almond, Coconut
f17, f24, f84, f92, Hazel nut, Shrimp, Kiwi, Banana
f201, f202, f203, f256, Pecan nut, Cashew nut, Pistachio, Walnut
f25, f214, f216, f218, Tomato, Spinach, Cabbage, Paprika
f25, f45, f47, f48, f85, Tomato, Yeast, Garlic, Onion, Celery
f26, f27, f83, Pork, Beef, Chicken
f265, f266, f267, f268, Caraway, Mace, Cardamom, Clove
f269, f219, f270, f271, Basil, Fennel seed, Ginger, Anise
f272, f274, f273, f275, Tarragon, Marjoram, Thyme, Lovage
f3, f205, f206, f254, Cod, Herring, Mackerel, Plaice
f3, f24, f37, f40, f41, Fish, Shrimp, Blue mussel, Tuna, Salmon
f33, f49, f92, f95, Orange, Apple, Banana, Peach
f4, f5, f6, f9, Wheat, Rye, Barley, Rice
f4, f7, f8, f10, f11, Wheat, Oat, Maize, Sesame seed, Buckwheat
f44, f94, f208, f210, Strawberrry, Pear, Lemon, Pineapple
f49, f92, f94, f95, Apple, Banana, Pear, Peach
f84, f87, f92, f95, f210, Kiwi, Melon, Banana, Peach, Pineapple
Regulatory Status:
ASR
FDA Cleared
This item is not returnable. View return policy
This item is not returnable. View return policy

Measure specifc IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.

Identify specific IgE antibodies to broad categories of allergens with ImmunoCAP™ Food Allergen Mixes, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to specific allergens. Qualitative measurement of circulating IgE antibodies to allergen mixes provide an assessment of sensitization to group of related allergens. Reflex testing to the individual whole allergens that make up an allergen mix is generally recommended to identify the allergen specific IgE.

ImmunoCAP Specific IgE Mixes offers the laboratory:
  • Detection limit ≥ 0.35 kUA/L
  • Qualitative results
  • Ability to detect specific IgE to multiple allergens in one test
  • Excellent consistency over time, and between countries, systems, labs and persons

Expected test values
Test results for ImmunoCAP Allergen Mixes are derived from multiple allergens and are reported as qualitative values (positive or negative). A cut-off value of 0.35 kUA/L is recommended for ImmunoCAP multiple allergens. Values between limit of quantification and 0.35 kUA/L may represent very low levels of IgE antibodies. Values ≥0.35 kUA/L indicate specific IgE antibodies to one or more of the allergens coupled to ImmunoCAP multiple allergen. Reinvestigation with appropriate ImmunoCAP single allergen is recommended when there is a need to further identify and obtain a quantitative result for the specific allergen(s). A value below 0.35 kUA/L indicates undetectable or very low levels of allergen specific IgE antibodies towards all of the allergens bound to ImmunoCAP multiple allergen but deviations from results obtained with single ImmunoCAP allergens may occur. The interpretation of results obtained with ImmunoCAP multiple allergen cannot be compared with the results with ImmunoCAP single allergen. The degree of positivity of ImmunoCAP multiple allergen cannot be considered the cumulative degree of positivity of the respective ImmunoCAP single allergen.

Specimen collection and preparation
  • Serum and plasma (EDTA or heparin) samples from venous or capillary blood can be used
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2-8°C up to one week, otherwise store at -20°C
  • Avoid repeated freezing and thawing

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:

  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:

  • Severe symptoms including asthma exacerbations—a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.

TRUSTED_SUSTAINABILITY

Specifications

Regulatory Status FDA Cleared
Code Type Food
Common Name f33, f49, f92, f95, Orange, Apple, Banana, Peach
Quantity 10 Tests
Code fx15
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.