Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ImmunoCAP™ Fruit and Vegetable Allergens

Catalog No. 14412901P
Change view
Click to view available options
Quantity:
10 Tests
16 Tests
Code:
Ff294
Rf211
Rf217
Rf261
Rf262
Rf288
Rf289
Rf291
Rf293
Rf301
Rf302
Rf306
Rf319
Rf322
Rf328
Rf329
Rf341
Rf342
Rf343
f208
f209
f210
f214
f215
f216
f225
f237
f242
f244
f25
f255
f259
f260
f31
f33
f35
f44
f47
f48
f49
f51
f54
f84
f85
f87
f91
f92
f94
f95
f96
Common Name:
Apple
Apricot
Asparagus
Aubergine, eggplant
Avocado
Bamboo shoot
Banana
Beetroot
Blackberry
Blueberry
Broccoli
Brussel sprouts
Cabbage
Carrot
Cauliflower
Celery
Cherry
Cranberry
Cucumber
Date
Fig
Garlic
Grape
Grapefruit
Kiwi
Lemon
Lettuce
Lime
Mandarin
Mango
Melon
Olive (black, fresh)
Onion
Orange
Papaya
Passion fruit
Peach
Pear
Persimmon
Pineapple
Plum
Potato
Pumpkin
Raspberry
Red currant
Spinach
Strawberry
Sweet potato
Tomato
Watermelon
50 product options available for selection
Product selection table with 50 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Code Common Name
14-412-901P 16 Tests f33 Orange
14-418-201P 16 Tests f25 Tomato
14-421-001P 16 Tests f31 Carrot
14-421-101P 16 Tests f35 Potato
14-421-601P 16 Tests f44 Strawberry
14-421-901P 16 Tests f48 Onion
14-421-801P 16 Tests f47 Garlic
14-422-001P 16 Tests f49 Apple
14-429-601P 10 Tests f85 Celery
14-429-501P 10 Tests f84 Kiwi
14-429-801P 10 Tests f87 Melon
14-433-401P 10 Tests f92 Banana
14-433-601P 10 Tests f94 Pear
14-433-101P 10 Tests f91 Mango
14-433-701P 10 Tests f95 Peach
14-434-601P 10 Tests f96 Avocado
14-443-101P 16 Tests f209 Grapefruit
14-443-201P 16 Tests f214 Spinach
14-443-301P 16 Tests f225 Pumpkin
14-445-501P 16 Tests f51 Bamboo shoot
14-445-801P 16 Tests f54 Sweet potato
14-480-901P 10 Tests f259 Grape
14-481-001P 10 Tests f210 Pineapple
14-481-101P 10 Tests f242 Cherry
14-481-201P 10 Tests f237 Apricot
14-481-301P 10 Tests f215 Lettuce
14-481-401P 10 Tests f216 Cabbage
14-482-901P 10 Tests f208 Lemon
14-483-901P 10 Tests f244 Cucumber
14-484-101P 10 Tests f260 Broccoli
14-484-001P 10 Tests f255 Plum
14-504-710P 10 Tests Rf262 Aubergine, eggplant
14-504-610P 10 Tests Rf261 Asparagus
14-505-110P 10 Tests Rf211 Blackberry
14-505-210P 10 Tests Rf288 Blueberry
14-506-010P 10 Tests Rf291 Cauliflower
14-505-410P 10 Tests Rf217 Brussel sprouts
14-506-810P 10 Tests Rf289 Date
14-508-910P 10 Tests Rf293 Papaya
14-509-110P 10 Tests Ff294 Passion fruit
14-516-110P 10 Tests Rf301 Persimmon
14-516-010P 10 Tests Rf302 Mandarin
14-518-810P 10 Tests Rf306 Lime
14-521-610P 10 Tests Rf319 Beetroot
14-524-410P 10 Tests Rf328 Fig
14-524-510P 10 Tests Rf329 Watermelon
14-526-910P 10 Tests Rf341 Cranberry
14-527-910P 10 Tests Rf342 Olive (black, fresh)
14-528-010P 10 Tests Rf343 Raspberry
14-522-110P 10 Tests Rf322 Red currant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 50 options available.
50 options
This item is not returnable. View return policy
Catalog No. 14-412-901P Supplier Thermo Scientific™ Supplier No. 14412901
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measure specifc IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.

Quantify specific IgE antibodies with ImmunoCAP™ Fruit and Vegetable Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.

ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.
TRUSTED_SUSTAINABILITY

Specifications

Scientific Name Citrus sinensis
Regulatory Status FDA Cleared
Code Type Fruit/Vegetable
Common Name Orange
Quantity 16 Tests
Code f33
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.