Learn More
Thermo Scientific™ ImmunoCAP™ Total IgE Reagents for Phadia™ 5000/2500
Catalog No. p-4373947
Click to view available options
:
ImmunoCAP Total IgE Control LMH; Size: 3 x 2 x 4
Specific IgE Curve Control Strip; Size: 5 x 3 pairs
Total IgE Anti-IgE ImmunoCAP; Size: 16
Total IgE Calibrator Strip; Size: 5 x 1 curve
Total IgE Conjugate 100; Size: 6 x 100
Total IgE Conjugate 400; Size: 6 x 400
Total IgE Curve Control Strip; Size: 5 x 3 pairs
This item is not returnable. View return policy
This item is not returnable. View return policy
Measures the quantitative amount of total IgE in human serum or plasma
TRUSTED_SUSTAINABILITY
Specifications
Product Type Specific IgE curve control strip
For Use With (Equipment) Phadia™ 5000/2500
Quantity 5 x 3 Pair
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.