Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ImmunoCAP™ Tree Allergen Mixes

Catalog No. 14420101P
Change view
Click to view available options
Quantity:
10 Tests
16 Tests
Code:
Rtx10
tx1
tx4
tx5
tx6
tx7
tx8
tx9
Common Name:
t1, t3, t4, t7, t11, Acer negundo, Betula verrucosa, Corylus avellana, Quercus alba, Platanus acerifolia
t1, t3, t5, t7, t10, Acer negundo, Betula verrucosa, Fagus grandifolia, Quercus alba, Juglans californica
t1, t3, t7, t8, t10, Acer negundo, Betula verrucosa, Quercus alba, Ulmus americana, Juglans californica
t2, t3, t4, t15, Alnus incana, Betula verrucosa, Corylus avellana, Fraxinus americana
t2, t3, t4, t7, t12, Alnus incana, Betula verrucosa, Corylus avellana, Quercus alba, Salix caprea
t2, t4, t8, t12, t14, Alnus incana, Corylus avellana, Ulmus americana, Salix caprea, Populus deltoides
t7, t8, t11, t12, t14, Quercus alba, Ulmus americana, Platanus acerifolia, Salix caprea, Populus deltoides
t9, t12, t16, t18, t19, t21, Olea europaea, Salix caprea, Pinus strobus, Eucalyptus spp., Acacia longifolia, Melaleuca leucadendron
Regulatory Status:
ASR
FDA Cleared
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Code Common Name Regulatory Status
14-420-101P 16 Tests tx5 t2, t4, t8, t12, t14, Alnus incana, Corylus avellana, Ulmus americana, Salix caprea, Populus deltoides FDA Cleared
14-419-901P 16 Tests tx1 t1, t3, t7, t8, t10, Acer negundo, Betula verrucosa, Quercus alba, Ulmus americana, Juglans californica FDA Cleared
14-420-001P 16 Tests tx4 t7, t8, t11, t12, t14, Quercus alba, Ulmus americana, Platanus acerifolia, Salix caprea, Populus deltoides FDA Cleared
14-420-201P 16 Tests tx6 t1, t3, t5, t7, t10, Acer negundo, Betula verrucosa, Fagus grandifolia, Quercus alba, Juglans californica FDA Cleared
14-427-301P 16 Tests tx8 t1, t3, t4, t7, t11, Acer negundo, Betula verrucosa, Corylus avellana, Quercus alba, Platanus acerifolia FDA Cleared
14-427-201P 16 Tests tx7 t9, t12, t16, t18, t19, t21, Olea europaea, Salix caprea, Pinus strobus, Eucalyptus spp., Acacia longifolia, Melaleuca leucadendron FDA Cleared
14-427-401P 16 Tests tx9 t2, t3, t4, t7, t12, Alnus incana, Betula verrucosa, Corylus avellana, Quercus alba, Salix caprea FDA Cleared
14-518-310P 10 Tests Rtx10 t2, t3, t4, t15, Alnus incana, Betula verrucosa, Corylus avellana, Fraxinus americana ASR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 14-420-101P Supplier Thermo Scientific™ Supplier No. 14420101
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measure specifc IgE antibodies using Thermo Scientific™ ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.

Identify specific IgE antibodies to broad categories of allergens with ImmunoCAP™ Tree Allergen Mixes, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to specific allergens. Qualitative measurement of circulating IgE antibodies to allergen mixes provide an assessment of sensitization to group of related allergens. Reflex testing to the individual whole allergens that make up an allergen mix is generally recommended to identify the allergen specific IgE.

ImmunoCAP Specific IgE Mixes offers the laboratory:
  • Detection limit ≥ 0.35 kUA/L
  • Qualitative results
  • Ability to detect specific IgE to multiple allergens in one test
  • Excellent consistency over time, and between countries, systems, labs and persons

Expected test values
Test results for ImmunoCAP Allergen Mixes are derived from multiple allergens and are reported as qualitative values (positive or negative). A cut-off value of 0.35 kUA/L is recommended for ImmunoCAP multiple allergens. Values between limit of quantification and 0.35 kUA/L may represent very low levels of IgE antibodies. Values ≥0.35 kUA/L indicate specific IgE antibodies to one or more of the allergens coupled to ImmunoCAP multiple allergen. Reinvestigation with appropriate ImmunoCAP single allergen is recommended when there is a need to further identify and obtain a quantitative result for the specific allergen(s). A value below 0.35 kUA/L indicates undetectable or very low levels of allergen specific IgE antibodies towards all of the allergens bound to ImmunoCAP multiple allergen but deviations from results obtained with single ImmunoCAP allergens may occur. The interpretation of results obtained with ImmunoCAP multiple allergen cannot be compared with the results with ImmunoCAP single allergen. The degree of positivity of ImmunoCAP multiple allergen cannot be considered the cumulative degree of positivity of the respective ImmunoCAP single allergen.

Specimen collection and preparation
  • Serum and plasma (EDTA or heparin) samples from venous or capillary blood can be used
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2-8°C up to one week, otherwise store at -20°C
  • Avoid repeated freezing and thawing

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:

  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:

  • Severe symptoms including asthma exacerbations—a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.

TRUSTED_SUSTAINABILITY

Specifications

Regulatory Status FDA Cleared
Code Type Tree
Common Name t2, t4, t8, t12, t14, Alnus incana, Corylus avellana, Ulmus americana, Salix caprea, Populus deltoides
Quantity 16 Tests
Code tx5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.