Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ImmunoCAP™ Tree Pollen Allergens

Catalog No. 14482301P
Change view
Click to view available options
Quantity:
10 Tests
16 Tests
Code:
Rt201
Rt205
Rt206
Rt209
Rt211
Rt213
Rt214
Rt217
Rt218
Rt37
Rt41
Rt44
Rt45
Rt55
Rt56
Rt57
Rt71
t1
t10
t11
t12
t14
t15
t16
t17
t18
t19
t2
t20
t208
t21
t210
t212
t22
t222
t23
t3
t4
t5
t6
t7
t70
t72
t73
t8
t9
Common Name:
Acacia
American Beech
Australian Pine
Bald Cypress
Bayberry
Box Elder
Cedar
Cedar Elm
Chestnut
Common Silver Birch
Cottonwood
Cypress
Date
Elder
Elm
Eucalyptus, Gum-tree
Grey Alder
Hackberry
Hazel
Horn Beam
Italian/Mediterranean/Funeral Cypress
Japanese Cedar
Linden
Maple Leaf Sycamore, London Plane
Melaleuca, Cajeput-tree
Mesquite
Mountain Juniper
Mulberry
Oak
Olive
Pecan, Hickory
Peppertree
Pine
Privet
Queen Palm
Red Cedar
Red Mulberry
Scotch Broom
Spruce
Sweet Gum
Virgina Live Oak
Walnut
White Ash
White Hickory
White Pine
Willow
46 product options available for selection
Product selection table with 46 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Code Common Name
14-482-301P 10 Tests t208 Linden
14-410-201P 16 Tests t3 Common Silver Birch
14-411-801P 16 Tests t17 Japanese Cedar
14-414-901P 16 Tests t7 Oak
14-414-701P 16 Tests t4 Hazel
14-414-801P 16 Tests t5 American Beech
14-414-601P 16 Tests t2 Grey Alder
14-415-001P 16 Tests t9 Olive
14-415-101P 16 Tests t12 Willow
14-416-101P 16 Tests t1 Box Elder
14-422-501P 16 Tests t6 Mountain Juniper
14-422-601P 16 Tests t8 Elm
14-422-901P 16 Tests t14 Cottonwood
14-422-701P 16 Tests t10 Walnut
14-423-001P 16 Tests t15 White Ash
14-423-201P 16 Tests t18 Eucalyptus, Gum-tree
14-423-301P 16 Tests t19 Acacia
14-423-101P 16 Tests t16 White Pine
14-422-801P 16 Tests t11 Maple Leaf Sycamore, London Plane
14-423-401P 16 Tests t20 Mesquite
14-423-501P 16 Tests t21 Melaleuca, Cajeput-tree
14-423-601P 16 Tests t22 Pecan, Hickory
14-423-701P 16 Tests t23 Italian/Mediterranean/Funeral Cypress
14-432-501P 10 Tests t70 Mulberry
14-437-301P 10 Tests t72 Queen Palm
14-437-401P 10 Tests t73 Australian Pine
14-483-401P 10 Tests t210 Privet
14-494-210P 10 Tests Rt41 White Hickory
14-494-110P 10 Tests Rt37 Bald Cypress
14-494-310P 10 Tests Rt44 Hackberry
14-494-410P 10 Tests Rt45 Cedar Elm
14-494-610P 10 Tests Rt55 Scotch Broom
14-494-710P 10 Tests Rt56 Bayberry
14-494-810P 10 Tests Rt57 Red Cedar
14-494-910P 10 Tests Rt71 Red Mulberry
14-500-210P 10 Tests Rt205 Elder
14-500-510P 10 Tests Rt201 Spruce
14-514-010P 10 Tests Rt206 Chestnut
14-514-810P 10 Tests Rt209 Horn Beam
14-515-510P 10 Tests Rt211 Sweet Gum
14-516-710P 10 Tests Rt217 Peppertree
14-518-501P 10 Tests t212 Cedar
14-520-810P 10 Tests Rt213 Pine
14-521-810P 10 Tests Rt214 Date
14-525-210P 10 Tests Rt218 Virgina Live Oak
14-530-901P 10 Tests t222 Cypress
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 46 options available.
46 options
This item is not returnable. View return policy
Catalog No. 14-482-301P Supplier Thermo Scientific™ Supplier No. 14482301
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measure specifc IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.

Quantify specific IgE antibodies with ImmunoCAP™ Tree Pollen Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.

ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.
TRUSTED_SUSTAINABILITY

Specifications

Scientific Name Tilia cordata
Regulatory Status FDA Cleared
Code Type Tree
Common Name Linden
Quantity 10 Tests
Code t208
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.