Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Immunostics Hema-Screen SPECIFIC™ iFOBT Complete Test Kit

Catalog No. 23040035 Shop All Immunostics Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-040-035 25 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-040-035 Supplier Immunostics Supplier No. HSGD25
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

25 cassettes, 25 buffer tubes, and 35 postal-approved mailing envelopes each containing one Devel-A-Tab slide, two collection devices, two applicators and patient instructions

Use for the rapid and qualitative determination of human blood in fecal sample. Innovative immunochemical collection system uses a single iFOBT cassette and a single patented Devel-A-Tab™ sampling card to detect human hemoglobin [Hb (globin)] in fecal stool specimens.

  • No dietary restrictions or interfering substances, encouraging patient compliance
  • Easy-to-use, take-home collection system; simply use an applicator and Devel-A-Tab collection card to obtain two consecutive specimens
  • Specimens are sealed in postal-approved envelopes and mailed to physician's office or testing lab
  • In-office CLIA waived procedure with test results in five minutes
  • Sensitivity level of 50μg Hb/g feces with 100% specificity for human hemoglobin
  • Recommended for use in routine physical examinations, hospital monitoring for bleeding from in-patient population, and for screening for colorectal cancer or gastrointestinal bleeding

Intended for professional and laboratory use only

Order Info

Up to 30-day specimen stability at room temperature

US Patent 7,427,505. European EP 1 619 502 B1.

Specifications

Clia Complexity Waived
Type iFOBT Complete Test Kit
For Use With (Application) Determination of Human hemoglobin in feces, a vital tool in the diagnosis and therapy of gastrointestinal disorders
Includes 25 Cassettes, 25 Buffer tubes, 35 mailing envelopes containing 1 DEVEL-A-TAB™ slide, 2 collection tissues, 2 applicator sticks, patient instructions and product insert, procedure card
Quantity 25 Pack
Test Time 5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.