Learn More
Immunostics Hema-Screen™ STAT FOBT Test
Use for the rapid, convenient and qualitative detection of asymptomatic occult bleeding.
Supplier: Immunostics HSTAT50
Includes: Laboratory Pack: instructions for use, single slides with Performance Standards, developing solution and applicator sticks
Description
- Self-contained test: no separate developing solution required
- Recommended use in routine office practice, routine hospital testing, mass screening programs for colorectal cancer and in testing of postoperative patients and newborn infants
- Slides can be prepared at patient's bedside and placed in sealed envelope
Specifications
Waived | |
Detection of fecal occult Blood | |
50 Pack |
Hema Screen™ Stat FOBT Test | |
50 Two hole test slides with on-slide control and built in developer, 50 applicator, 1 product insert |