Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IMMY Myco DDR™ Mycobacterial Digestion/ Decontamination Kits and Components

Click to view available options
Type:
NaOH/NALC 2.5% Bulk
NaOH/NALC 3.0% Bulk
Neutralization Buffer B
Resuspension Buffer C
Description
- Culture results obtained faster
- Fewer specimens to reprocess
- Reduced risk of cross-contamination
- Easy to process specimens containing blood
- Reduces sample splitting workload
- Eliminates user variability, reducing the chances for technical error and allowing samples to be easily processed
- Advanced buffer reduces the number of samples that need to be split
- Cleaner smears
- Visual result confirmation
Specifications
Specifications
| Type | Neutralization Buffer B |
| For Use With (Application) | Mycobacterial testing |
| Quantity | 60 x 30 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction