Learn More
IMP1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303423A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat
IGF2BP1 is a member of the insulin-like growth factor 2 mRNA-binding protein family. The protein contains four K homology domains and two RNA recognition motifs. It functions by binding to the mRNAs of certain genes, including insulin-like growth factor 2, beta-actin and beta-transducin repeat-containing protein, and regulating their translation. Two transcript variants encoding different isoforms have been found for this gene.Specifications
IMP1 | |
Polyclonal | |
Unconjugated | |
IGF2BP1 | |
IGF II mRNA binding protein 1; IGF-II mRNA-binding protein 1; IGF2 mRNA-binding protein 1; VICKZ family member 1; ZBP-1; coding region determit-binding protein; insulin-like growth factor 2 mRNA-binding protein 1; zip code-binding protein 1; zipcode-binding protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10642 | |
4° C | |
Liquid |
Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9NZI8 | |
IGF2BP1 | |
Between 150 and 200 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.