Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IMP2/IGF2BP2 Antibody (SR2106), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | IMP2/IGF2BP2 |
---|---|
Clone | SR2106 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
IMP2/IGF2BP2 Monoclonal antibody specifically detects IMP2/IGF2BP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
IMP2/IGF2BP2 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Hepatocellular carcinoma autoantigen p62, IGF-II mRNA-binding protein 2, IMP-2IGF2 mRNA-binding protein 2, IMP2VICKZ family member 2, insulin-like growth factor 2 mRNA binding protein 2, insulin-like growth factor 2 mRNA-binding protein 2, p62, VICKZ2 | |
A synthesized peptide derived from human IMP2/IGF2BP2 (Uniprot #: Q9Y6M1) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2106 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Chromatin Research, Immune System Diseases, Immunology, Transcription Factors and Regulators | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
10644 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title