Learn More
Invitrogen™ IMP3 Polyclonal Antibody

Description
Immunogen sequence: DFLNWEVTDH NLHELRVLRR YRLQRREDYT RYNQLSRAVR ELARRLRDLP ERDQFRVRAS AALLDKLYAL GLVPT Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 96%.
Specifications
Specifications
| Antigen | IMP3 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.20 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Imp3 |
| Gene Accession No. | Q9NV31 |
| Gene Alias | 1190002L16Rik; AI256594; BRMS2; C15orf12; cancer/testis antigen 98; CT98; DKFZp586L0118; FLJ10968; hKOC; IGF2 mRNA-binding protein 3; IGF2BP3; IGF-II mRNA-binding protein 3; IMP3; IMP-3; IMP3, U3 small nucleolar ribonucleoprotein; IMP3, U3 small nucleolar ribonucleoprotein, homolog; IMP3, U3 small nucleolar ribonucleoprotein, homolog (yeast); insulin like growth factor 2 mRNA binding protein 3; insulin-l; insulin-like growth factor 2 mRNA binding protein 3; insulin-like growth factor 2 mRNA-binding protein 3; KH domain containing protein overexpressed in cancer; KH domain-containing protein overexpressed in cancer; KOC; KOC1; MRPS4; OTTHUMP00000183524; RGD1306825; U3 small nucleolar ribonucleoprotein protein IMP3; U3 snoRNP protein 3 homolog; U3 snoRNP protein IMP3; VICKZ family member 3; VICKZ3 |
| Gene Symbols | Imp3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.