Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IMP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50
Specifications
Antigen | IMP3 |
---|---|
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5111512
|
Invitrogen™
PA5111512 |
100 μL |
Each for $589.50
|
|
|||||
PIPA5111518
|
Invitrogen™
PA5111518 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: TRGSLELCDF VTASSFCRRR LPTVLLKLRM AQHLQAAVAF VEQGHVRVGP DVVTDPAFLV TRSMEDFVTW VDSSKIKRHV LEYNEERDDF DLE.
This gene encodes the human homolog of the yeast Imp3 protein. The protein localizes to the nucleoli and interacts with the U3 snoRNP complex. The protein contains an S4 domain.Specifications
IMP3 | |
Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NV31 | |
55272 | |
Recombinant Protein corresonding to Human IMP3. Recombinant protein control fragment (Product #RP-105162). | |
Antibody |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
Imp3 | |
1190002L16Rik; AI256594; BRMS2; C15orf12; cancer/testis antigen 98; CT98; DKFZp586L0118; FLJ10968; hKOC; IGF2 mRNA-binding protein 3; IGF2BP3; IGF-II mRNA-binding protein 3; IMP3; IMP-3; IMP3, U3 small nucleolar ribonucleoprotein; IMP3, U3 small nucleolar ribonucleoprotein, homolog; IMP3, U3 small nucleolar ribonucleoprotein, homolog (yeast); insulin like growth factor 2 mRNA binding protein 3; insulin-l; insulin-like growth factor 2 mRNA binding protein 3; insulin-like growth factor 2 mRNA-binding protein 3; KH domain containing protein overexpressed in cancer; KH domain-containing protein overexpressed in cancer; KOC; KOC1; MRPS4; OTTHUMP00000183524; RGD1306825; U3 small nucleolar ribonucleoprotein protein IMP3; U3 snoRNP protein 3 homolog; U3 snoRNP protein IMP3; VICKZ family member 3; VICKZ3 | |
Imp3 | |
Primary | |
Antigen affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title