Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IMP3 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501565103
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-5103 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-5103 Supplier Bethyl Laboratories, Inc Supplier No. A303426AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Insulin-like growth factor-II messenger RNA (mRNA)-binding protein-3 (IMP-3), also known as K homology domain-containing protein overexpressed in cancer (KOC) and L523S, is a member of the insulin-like growth factor-II mRNA-binding protein family and is expressed during embryogenesis and in some maligcies. IMP-3 is expressed in maligt melanoma but not in benign nevi, even when dysplastic features are present; IMP-3 is expressed in a significantly higher proportion of melanomas than Spitz nevi; and IMP-3 is expressed in metastatic melanomas significantly more than in thin melanomas. IMP-3 appears to be involved in the progression of maligt melanoma and may play an important role in the regulation of the biologic behavior of this tumor.

Specifications

Antigen IMP3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene IGF2BP3
Gene Accession No. O00425, Q9CPN8
Gene Alias CT98, IMP-3, IMP3, KOC1, VICKZ3, IGF II mRNA binding protein 3, IGF-II mRNA-binding protein 3, IGF2 mRNA-binding protein 3, KH domain containing protein overexpressed in cancer, KH domain-containing protein overexpressed in cancer, VICKZ family member 3, cancer/testis antigen 98, hKOC, insulin-l
Gene Symbols IGF2BP3
Host Species Rabbit
Immunogen Between 529 and 579
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10643, 140488
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.