Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IMPA1 Antibody (2G5), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259416MFV500
Description
IMPA1 Monoclonal antibody specifically detects IMPA1 in Human samples. It is validated for Flow CytometrySpecifications
IMPA1 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
2G5 | |
Flow Cytometry | |
EC 3.1.3.25, IMP, IMP 1, IMPA, IMPase 1, inositol monophosphatase 1, inositol(myo)-1(or 4)-monophosphatase 1, Inositol-1(or 4)-monophosphatase 1, Lithium-sensitive myo-inositol monophosphatase A1 | |
Recombinant human IMPA1 (1-277aa) purified from E. coli | |
0.1 mL | |
Lipid and Metabolism | |
3612 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction