Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IMPA1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297107MFV450
Description
IMPA1 Polyclonal antibody specifically detects IMPA1 in Human samples. It is validated for ELISASpecifications
IMPA1 | |
Polyclonal | |
ELISA | |
EC 3.1.3.25, IMP, IMP 1, IMPA, IMPase 1, inositol monophosphatase 1, inositol(myo)-1(or 4)-monophosphatase 1, Inositol-1(or 4)-monophosphatase 1, Lithium-sensitive myo-inositol monophosphatase A1 | |
Produced in rabbits immunized with purified, recombinant Human IMPA1 (Accession#: P29218-1; Met1-Asp277) | |
0.1 mL | |
Lipid and Metabolism | |
3612 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction