Learn More
Invitrogen™ IMPA1 Monoclonal Antibody (1E6-F11)

Description
Peptide Sequence: MADPWQECMD YAVTLARQAG EVVCEAIKNE MNVMLKSSPV DLVTATDQKV EKMLISSIKE KYPSHSFIGE ESVAAGEKSI LTDNPTWIID PIDGTTNFVH RFPFVAVSIG FAVNKKIEFG VVYSCVEGKM YTARKGKGAF CNGQKLQVSQ QEDITKSLLV TELGSSRTPE TVRMVLSNME KLFCIPVHGI RSVGTAAVNM CLVATGGADA YYEMGIHCWD VAGAGIIVTE AGGVLMDVTG GPFDLMSRRV IAANNRILAE RIAKEIQVIP LQRDDED.
Specifications
Specifications
| Antigen | IMPA1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1E6-F11 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Impa1 |
| Gene Accession No. | P29218 |
| Gene Alias | 2610002K09Rik; 2900059K10Rik; AI325909; D-galactose 1-phosphate phosphatase; IMP; IMP 1; IMPA; IMPA1; Impa1-L; IMPase 1; inositol (myo)-1(or 4)-monophosphatase 1; inositol monophosphatase 1; inositol(myo)-1(or 4)-monophosphatase 1; inositol-1(or 4)-monophosphatase 1; Lithium-sensitive myo-inositol monophosphatase A1; myo-inositol monophosphatase 1; testicular tissue protein Li 94 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.