Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IMPA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556189
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556189 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556189 Supplier Invitrogen™ Supplier No. PA556189
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WEECFQAAVQ LALRAGQIIR KALTEEKRVS TKTSAADLVT ETDHLVEDLI ISELRERFPS HRFIAEEAAA SGAKCVLTHS PTWIIDPI Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.

This locus encodes an inositol monophosphatase. The encoded protein catalyzes the dephosphoylration of inositol monophosphate and plays an important role in phosphatidylinositol signaling. This locus may be associated with susceptibility to bipolar disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IMPA2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IMPA2
Gene Accession No. O14732
Gene Alias 2210415D20Rik; AI326924; AW259601; hypothetical protein LOC553595; IMP 2; IMP.18P; Impa2; IMPase 2; inosine monophosphatase 2; inositol (myo)-1(or 4)-monophosphatase 2; inositol monophosphatase 2; inositol monophosphatase 2 variant 1; inositol monophosphatase 2 variant 2; inositol(myo)-1(or 4)-monophosphatase 2; inositol-1(or 4)-monophosphatase 2; myo-inositol monophosphatase 2; myo-inositol monophosphatase A2; zgc:110201
Gene Symbols IMPA2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IMPA2. Recombinant protein control fragment (Product #RP-91745).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3613
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.