Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IMPACT Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP142678AF405
Description
IMPACT Polyclonal antibody specifically detects IMPACT in Human samples. It is validated for Western BlotSpecifications
IMPACT | |
Polyclonal | |
Western Blot | |
Impact homolog (mouse), Imprinted and ancient gene protein homolog, MGC33718, protein IMPACT | |
The immunogen for this product maps to a region between residue 270 and 320 of human imprinted and ancient homolog using the numbering given in entry NP_060909.1 (GeneID 55364). | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
55364 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction