Learn More
GSC Go Science Crazy Impact Car
Catalog No. s24646 Shop All GSC Products
Great tool for the study of force and motion, impacts and collisions
The impact car demonstrates force of an impact in a collision against a stationary object. The car is fitted with a spring scale and slide that is displaced on impact. The slide remains displaced until reset so measurements can be read. The top of the car can be removed to add weights. Provided with manual.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.