Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IMPACT Polyclonal Antibody

IMPACT Polyclonal Antibody
Supplier: Thermo Scientific PA559409
Description
IMPACT Polyclonal Antibody for Western Blot, IHC

Specifications
IMPACT | |
Polyclonal | |
Unconjugated | |
IMPACT | |
E430016J11Rik; Impact; Impact homolog; impact RWD domain protein; impact, RWD domain protein; imprinted and ancient gene protein; Imprinted and ancient gene protein homolog; protein IMPACT; RWD domain containing 5; RWDD5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55364 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2X3 | |
IMPACT | |
Recombinant protein corresponding to Human IMPACT. Recombinant protein control fragment (Product #RP-98247). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction