Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ IMPACT, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABN1444MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN1444MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1444MI Supplier MilliporeSigma™ Supplier No. ABN1444

Rabbit Polyclonal Antibody

Protein IMPACT (UniProt Q9P2X3; also known as Imprinted and ancient gene protein homolog) is encoded by the IMPACT gene (Gene ID 55364) in human. IMPACT is a cytoplasmic protein preferentially expressed in neurons. IMPACT is highly abundant in the hypothalamus and in scattered neurons in other areas, such as the hippocampal interneurons. IMPACT and its yeast ortholog Yih1 contain an RWD domain (a.a. 14-116 of human and mouse IMPACT) that interacts with GCN1 and competes against GCN2 for GCN1 binding. IMPACT and Yih1 overexpression in mouse embryonic fibroblasts (MEFs) and yeasts inhibits GCN2 activation induction. IMPACT is a developmentally regulated protein that promotes neuritogenesis, while GCN2 strongly inhibits spontaneous neurite outgrowth. Increased abundance of IMPACT in differentiated neuronal cells enhances translation initiation by preventing GCN2 signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IMPACT
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4) 150mM NaCl with 0.05% Sodium Azide.
Gene IMPACT
Gene Accession No. Q9P2X3
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the C-terminal sequence of human IMPACT.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_060909
Test Specificity This polyclonal antibody was raised against the C-terminal end sequence present in both spliced isoforms of human IMPACT reported by UniProt (Q9P2X3).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.