Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Three Quarter Finger Anti-Impact Nylon Gloves with VEP Padding

Catalog No. 19039264
Click to view available options
Size:
Large
Hand:
Left
Specific
This item is not returnable. View return policy
This item is not returnable. View return policy

VEP padding with a suede leather cover on the palm provides protection from impact and abrasion. Nylon fabric on the back ensures breathability and comfort.

  • Nylon fabric ensures breathability and comfort
  • Contoured VEP padding in the palm provides protection from impact and shock
  • 3/4 finger design leaves finger tips open to for tactile feedback and mobility
  • Suede leather cover offers abrasion protection

Specifications

Back Material Nylon Lycra™
Color Blue, Gray
Hand Specific
Material Suede
Product Type Anti-impact Gloves
Size Large
Finger Style Three Quarter
For Use With (Application) Construction, Assembly, Manufacturing, Maintenance, and Warehousing
Glove Cuff Style Open
Series 505-10
Length (English) 9 in.
Length (Metric) 23 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.