Learn More
Impacto™ Air Belt Sport Plus Fire Retardant Back Support
Shop All Impacto Protective Products Products
Description
Impacto™ Air Belt Sports Plus
- Breathable cotton outer cover
- Unique patented inflatable air technology
- Multi-celled air chambers conform to the back when air is added to provide relief from back pain.
- Natural therapy and support
Specifications
Specifications
| Color | Blue |
| Closure Type | Hook and loop |
| Product Type | Back Support |
| Design | Lumbar support; Inflatable multi celled air chambers; Palm-grip, self contained aspirator with pressure release valve |
| Size | Small |
| For Use With (Application) | Providing back support |
| Material | Cotton |
| Model | Sport Plus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.