Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Air Belt Sport Plus Fire Retardant Back Support

Catalog No. 19072385 Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
X-Large
Material:
Cotton
Proban FR Cotton Shell, Foam Liner
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Material
19-072385 X-Large Cotton
19-072382 Small Cotton
19-072668 Medium Proban FR Cotton Shell, Foam Liner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-072385 Supplier Impacto™ Supplier No. 24BSPXL
Add to Cart
Add to Cart

Impacto™ Air Belt Sports Plus with Fire Retardant cover offer natural therapy to support the back and help alleviate pain and discomfort

Impacto™ Air Belt Sports Plus

  • Breathable cotton outer cover
  • Unique patented inflatable air technology
  • Multi-celled air chambers conform to the back when air is added to provide relief from back pain.
  • Natural therapy and support

Specifications

Color Blue
Closure Type Hook and loop
Product Type Back Support
Design Lumbar support; Inflatable multi celled air chambers; Palm-grip, self contained aspirator with pressure release valve
Size X-Large
For Use With (Application) Providing back support
Material Cotton
Model Sport Plus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.