Learn More
Impacto™ Air Belt Sport Plus Fire Retardant Back Support
Impacto™ Air Belt Sports Plus with Fire Retardant cover offer natural therapy to support the back and help alleviate pain and discomfort
Supplier:  Impacto™ SPFRL
 View more versions of this product
Catalog No. 19-072667
Only null left
Add to Cart
Description
Description
Impacto™ Air Belt Sports Plus FR
  • Fire retardant cotton outer cover for work around sparks
  • Unique patented inflatable air technology
  • Multi-celled air chambers conform to the back when air is added to provide relief from back pain
  • Natural therapy and support
Specifications
Specifications
Hook and loop
Lumbar support; Inflatable multi celled air chambers; Palm-grip, self contained aspirator with pressure release valve
Providing back support
Sport Plus
Back Support
Large
Proban FR Cotton Shell, Foam Liner
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.