Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Impacto™ Air Belt Sport Plus Fire Retardant Back Support
Shop All Impacto Protective Products Products
Click to view available options
Size:
Small
Medium
X-Large
Material:
Cotton
Proban FR Cotton Shell, Foam Liner
Description
Impacto™ Air Belt Sports Plus
- Breathable cotton outer cover
- Unique patented inflatable air technology
- Multi-celled air chambers conform to the back when air is added to provide relief from back pain.
- Natural therapy and support
Specifications
Specifications
Color | Blue |
Closure Type | Hook and loop |
Product Type | Back Support |
Design | Lumbar support; Inflatable multi celled air chambers; Palm-grip, self contained aspirator with pressure release valve |
Size | X-Large |
For Use With (Application) | Providing back support |
Material | Cotton |
Model | Sport Plus |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction