Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Full-Finger Anti-Impact Grain Leather Gloves

Catalog No. 1899983A
Click to view available options
Size:
Small
Medium
Large
X-Large
Hand:
Left
Right
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves are stitched with DuPont™ Aramid Fibers for durability and abrasion protection. VEP padding in the palm, fingers, and thumb provides users with impact protection.

  • Durable grain leather offers abrasion protection and ensures durability
  • Contoured VEP 1/8 in. padding in the palm, fingers, and thumb protects from impact
  • Full finger style for protection of the entire hand
  • 100% aramid fiber stitching increases the glove's durability
  • Elastic gathering at the wrist helps to prevent debris from getting inside of the glove
  • Safety suede leather cuff with outer seams to ensure user comfort
  • Protection from air and power tools in applications such as construction, and maintenance

Specifications

Cuff Material Suede Leather
Color Yellow
Hand Left
Material Grain Leather
Product Type Anti-impact Gloves
Size Small
Finger Style Full
For Use With (Application) Construction, Maintenance
Glove Cuff Color White
Series 615-20
Length (English) 7 in.
Length (Metric) 18 cm
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.