Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Anti-Vibration Nylon Gloves with Grain Leather Tool Grip

Catalog No. 19031969
Click to view available options
Size:
Small
Medium
Large
X-Large
Hand:
Left
Right
Specific
This item is not returnable. View return policy
This item is not returnable. View return policy

VEP padding in the palm, thumb webbing, and index finger protects users from cumulative trauma caused by constant tool gripping.

  • Fingerless leather glove with stretchy nylon back
  • Soft grain leather palm cover for longer wear
  • Impact and shock absorbing VEP padding in the palm, between the thumb, and on the index finger
  • Thumb web area is protected from cumulative trauma such as bruising from gripping tools
  • Provides protection from impact, vibration, and abrasion
  • Ideal for use with hand tools and power tools

Specifications

Back Material Nylon
Color Blue, Yellow
Hand Right
Material Grain Leather
Product Type Tool Gloves
Size Small
Finger Style Fingerless
For Use With (Application) Construction, DIY Home Improvement
Glove Cuff Style Open
Series 502-20
Length (English) 7 in.
Length (Metric) 18 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.