Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Full Finger Anti-Vibration Air Glove™

Catalog No. 19048712C
Click to view available options
:
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

These leather gloves are ideal for use with vibrating power tools in construction, or industrial settings. Patented Air Bubble technology increases protection in the palm, thumb, and fingers.

  • Maximum vibration protection
  • Individual air chambers ensure long wear
  • Consistent protection where padded
  • Durable urethane bladder construction won't bottom out
  • Flexible around tool handles
  • Meets anti-vibration glove standards

Specifications

Color Yellow
Hand Specific
Material Cowhide Leather
Product Type Anti-vibration Gloves
Size Large
Finger Style Full
For Use With (Application) Construction, Power Tools, and Manufacturing
Glove Cuff Style Open
Series AIR650
Length (English) 9 in.
Length (Metric) 23 cm
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.