Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Anti-Impact Pearl Leather Gloves with VEP Waffle Padding

Catalog No. 19812021
Click to view available options
:
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

VEP padding extends from the palm to the base of the fingers to protect from impact and shock. The half finger design provides tactile feedback and mobility.

  • Pearl leather with soft lining protects from abrasion
  • 3/16 in. VEP waffle padding protects from impact and shock
  • Half finger design leaves fingers open for dexterity and mobility
  • Adjustable hook and loop wrist closure
  • Protects hands from manual and power tools

Specifications

Color White
Hand Right
Material Pearl Leather
Product Type Anti-impact Gloves
Size Large
Finger Style Half
For Use With (Application) Driving, Construction, Assembly, and Manufacturing
Glove Cuff Style Hook and Loop
Series 460-30
Length (English) 9 in.
Length (Metric) 23 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.