Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Cush'N Step Anti-Fatigue Molded Insoles

Impacto™ Cush'N Step CUSHMOLD is Made of lightweight cellular cushioning open-cell foam with re-reinforced gel inserts in the heel

$47.18 - $47.94

Specifications

Color Blue
For Use With (Application) Sports shoes, work boots and work footwear
Material Polymeric Open-cell Foam
Style Molded
Product Type Molded Insole
View More Specs

Products 2
Catalog Number Mfr. No. Size Gender Price Quantity & Availability  
Catalog Number Mfr. No. Size Gender Price Quantity & Availability  
19-062960
Encompass
View Documents
Impacto™
CUSHMOLDA
5 to 6.5 Women's
Pack for $47.18
Case of 12 PK for $565.08
Only null left
Add to Cart
 
19-062962
Encompass
View Documents
Impacto™
CUSHMOLDB
7 to 8.5 Women's
Pack for $47.94
Only null left
Add to Cart
 
Description

Description

Impacto™ Cush'N Step Anti-Fatigue Insoles

  • Cellular cushioning open cell foam with antibacterial fabric cover
  • Molded design with GEL heel insert and arch support
  • Helps relieve foot, legs and back fatigue
  • Pain and fatigue relief when standing or walking on hard, cold surfaces for long periods
  • For use with closed shoes, boots and sports shoes

† Also available in case of 12 packs.

Specifications

Specifications

Blue
Polymeric Open-cell Foam
Molded Insole
Sports shoes, work boots and work footwear
Molded
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.